Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%
Catalog # APOSOR965038-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%
Catalog # APOSOR965038-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethyl 3-(2-Pyrazinyl)-3-oxopropanoate, 3-Oxo-3-(pyrazin-2-yl)propionic acid Ethyl ester
 :
 :194.19 g/mol
Storage Temperature:Ambient
CAS Number:62124-77-0
Formula:C₉H₁₀N₂O₃
MDL Number:MFCD04114374