Di-tert-butyl malonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:541-16-2
:
Formula:C₁₁H₂₀O₄
MDL Number:MFCD00008810
Boiling Pt:65…67 °C/1mmHg
Density:1.42 g/cm³ (25 °C)
Molecular Weight:216.28 g/mol
Flash Pt:89 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together