1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSOR29643-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Ethyl-3-methylpyrazole-5-carboxylic acid
:
:154.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:50920-65-5
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD00156147
Melting Pt:142…143 °C