7,4'-Dihydroxy-6-methoxyisoflavone-7-D-glucoside
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Glycitin, 3-(4-Hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
CAS Number:40246-10-4
:
Formula:C₂₂H₂₂O₁₀
MDL Number:MFCD00800711
Molecular Weight:446.41 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together