1-Benzyl-azetidine-2-carboxylic acid amide 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Benzylazetidine-2-carboxamide
CAS Number:40432-40-4
:
Formula:C₁₁H₁₄N₂O
MDL Number:MFCD06658337
Molecular Weight:190.242 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together