2-Methyl-1,3-benzothiazole-5-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Methylbenzo[d]thiazole-5-carboxylic acid
CAS Number:24851-69-2
:
Formula:C₉H₇NO₂S
MDL Number:MFCD00498510
Molecular Weight:193.222 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together