Adefovir dipivoxil
: Adefovir Dipivoxil, 9-(2[bis(Pivaloyloxymethoxy)phosphorylmethoxy]ethyl)adenine, 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine, ((((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphoryl)bis(oxy))bis(methylene) bis(2,2-dimethylpropanoate)
:142340-99-6
:
:C₂₀H₃₂N₅O₈P
:MFCD00869897
:501.48 g/mol
:100 °C
:GHS07
:Freezer
- Environmentally Preferable:
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Melting point:98 to 103 °C
- Titration with HClO4:98.0 to 102.0 %
Frequently Bought Together