Copper(II) acetylacetonate for synthesis, Sigma-Aldrich®
Catalog # 8.02712.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.2 g/l (20 °C)
- Melting Point:245 °C (decomposition)
- Molar Mass:261.76 g/mol
- Bulk Density:200 - 400 kg/m3
- Boiling Point:160 °C (13 hPa)
- Vapor Pressure:0.13 hPa (163 °C)
Specification Test Results
About this item
Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
:
:261.76 g/mol
EINECS:236-477-9
:GHS07
CAS Number:13395-16-9
Formula:C₁₀H₁₄CuO₄
MDL Number:MFCD00000016
Melting Pt:279…283 °C
:M-Clarity™ quality level = MQ200