EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester) ≥98.0% (by total nitrogen basis) cross-linking reagent
Supplier: TCI
Certificates
About this item
Synonyms: 6-Maleimidocaproic acid N-succinimidyl ester, N-Succinimidyl 6-Maleimidohexanoate, EMCS, 2,5-Dioxopyrrolidin-1-yl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate, N-(6-Maleimidocaproyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate, N-ε-malemidocaproyl-oxysuccinimide ester, N-(ε-Maleimidocaproyloxy)succinimide, 6-Maleimidohexanoic acid N-hydroxysuccinimide ester
CAS Number:55750-63-5
:
Formula:C₁₄H₁₆N₂O₆
MDL Number:MFCD00043043
Molecular Weight:308.29 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:69.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together