trans-1,4-Dichlorocyclohexane ≥99.0% (by GC)
Catalog # TCIAD1425-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:102.0 to 104.0 deg-C
Specification Test Results
About this item
:
:153.05 g/mol
CAS Number:16890-91-8
Formula:C₆H₁₀Cl₂
MDL Number:MFCD00070474
Boiling Pt:60°C