1,3-Cyclopentanediol (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAC2534-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
Specification Test Results
About this item
Synonyms: 1,3-Dihydroxycyclopentane
:
:102.13 g/mol
EINECS:261-880-1
CAS Number:59719-74-3
Formula:C₅H₁₀O₂
MDL Number:MFCD00001367
Boiling Pt:91°C