Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Colour Reference Solutions, set of colours Y, ampoules of 2 ml, Merck
Supelco® Colour Reference Solutions, set of colours Y, ampoules of 2 ml, Merck
Catalog # 83883-1SET-F
Supplier:  Merck
CAS Number:  
Supelco® Colour Reference Solutions, set of colours Y, ampoules of 2 ml, Merck
Catalog # 83883-1SET-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Ampoule, T/E
  • Pk:
    1 KIT
  • Description:
    Set of colours Y
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100

Specifications

About this item

The kit contains a set of seven yellow colour reference standards (identified sequentially from Y1 to Y7) and a 1% hydrogen chloride solution in water. The solutions are produced from two primary metal salt solutions of ferric(III) chloride and cobalt(II) chloride, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 2 ml ampoules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

They are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red colour range.

This is a set of the following color reference solutions according to PhEur:

Colour Reference Solution Y1 - 2 ml ampoule
Colour Reference Solution Y2 - 2 ml ampoule
Colour Reference Solution Y3 - 2 ml ampoule
Colour Reference Solution Y4 - 2 ml ampoule
Colour Reference Solution Y5 - 2 ml ampoule
Colour Reference Solution Y6 - 2 ml ampoule
Colour Reference Solution Y7 - 2 ml ampoule
Blind sample : Hydrochloric acid solution, 1% in water - 2 ml ampoule

Caution: Contains Cobalt(II) chloride.