Lithium tetraborate : lithium metaborate 35,3:64,7 (w/w), Spectroflux® 100D
Catalog # 97890.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):White
- Form:Powder
- Comment, Li:B Ratio:0.492 - 0.544
- Elemental Analysis, Boron B (Theoretical):22.08%
- Elemental Analysis, Lithium Li (Theoretical):11.95%
- Residue on Ignition:Max. 1.5% (30 mins, 900 °C)
- Impurity Content, Aluminium (Al):Max. 50 ppm
- Impurity Content, Calcium (Ca):Max. 50 ppm
- Impurity Content, Others, Each:Max. 15 ppm
- Impurity Content, Potassium (K):Max. 50 ppm
- Impurity Content, Silicon (Si):Max. 20 ppm
- Impurity Content, Sodium (Na):Max. 50 ppm
- Total Metal Impurities:Max. 100 ppm
Specification Test Results
About this item
:
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD01074946