Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, EPA 610 Polynuclear Aromatic Hydrocarbons Mix
  CRM48743
 :  Merck
 :  
Supelco® Organic Reference Standard, EPA 610 Polynuclear Aromatic Hydrocarbons Mix
  CRM48743
 :  Merck
 :  

 

  • Concentration:
    In methanol: methylene chloride (1:1) (varied concentration)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    EPA 610 Polynuclear aromatic hydrocarbons mix
  • Environmentally Preferable:
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H315 - H319 - H336 - H350 - H370 - H410
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    EPA 610 Polynuclear Aromatic Hydrocarbons Mixture
  • Flash point:
    11,0 °C - Closed cup

 

 

  • GHS signal: GHS02, GHS06, GHS08, GHS09
 : P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311