Paraformaldehyde 4% in PBS ready-to-use, fixative reagent
Catalog # 22023.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
- Pk Info:5×20 ml
Specifications
About this item
Synonyms: Polyoxymethylene
:
:GHS06
:GHS07
:GHS08
CAS Number:30525-89-4
Formula:(CH₂O)n
MDL Number:MFCD00133991
Melting Pt:123
Flash Pt:70
Merck Index:13,07096
:Packaged under argon.