Tin(IV) oxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide
CAS Number:18282-10-5
:
Formula:O₂Sn
MDL Number:MFCD00011244
Boiling Pt:1800…1900 °C (subl.)
Melting Pt:1630 °C
Density:6.95
Molecular Weight:150.71 g/mol
Merck Index:14,8778
Storage Temperature:Ambient
EINECS:242-159-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together