Bromocresol green, powder pH-indicator
Catalog # 200125B
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Transition range pH 3.8 - 5.4:Yellowish green to blue
- Appearance of solution:Passes test
- Transition range (according to ACS):Passes test
- Loss on drying (110°C):Max. 3.00 %
- Conforms to ACS:Passes test
Specification Test Results
About this item
Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
:
:698.02 g/mol
Melting Pt:217…218 °C
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
Merck Index:13,01370