Order Entry
Netherlands
ContactUsLinkComponent
Urea
Urea
Catalog # PROONIST-2152
CAS Number:  
undefined
Urea
Catalog # PROONIST-2152
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
 :
 :60.06 g/mol
Boiling Pt:197
Melting Pt:132.7
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Merck Index:13,09935