Acetyl chloride 99+%
: Acetyl chloride 1M in Dichloromethane
:75-36-5
:
:C₂H₃ClO
:MFCD00000719
:51 °C (760 mmHg)
:–112 °C
:1.1 g/cm³
:78.5 g/mol
:4 °C
:1717
:3 II
:13,00087
:200-865-6
:GHS02
:GHS05
:Ambient
- Environmentally Preferable:
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3875 to 1.3895 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤20 APHA
- Titration RCOCl:99.0 to 101.0 %
Frequently Bought Together