Oxalic acid dihydrate, Supelco®
: Merck
: Ethanedioic acid dihydrate
:6153-56-6
:
:HO₂CCO₂H·2H₂O
:MFCD00149102
:149…160 °C (1013 hPa)
:101
:1.65 g/cm³ (20 °C)
:126.07 g/mol
:13,06980
:M-Clarity™ quality level = MQ200
:Ambient
:205-634-3
:3261
:01-2119534576-33
:GHS05
:GHS07
- Environmentally Preferable:
- Solubility:102 g/l (20 °C)
- Melting Point:101 °C
- Molar Mass:126.07 g/mol
- Boiling Point:149 - 160 °C (decomposition)
- Vapor Pressure:21 hPa (50 °C)
- Flash Point:157 °C (decomposition)
- Density:1.65 g/cm3 (20 °C)
- pH:1 (10 g/l, H2O, 20 °C)
Frequently Bought Together