Sodium acetate trihydrate, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Acetic acid sodium salt trihydrate
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58
Density:1.4 g/cm³ (20 °C)
Molecular Weight:136.08 g/mol
Merck Index:13,08642
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:204-823-8
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:613 g/l (20 °C)
- Melting Point:58 °C
- Molar Mass:136.08 g/mol
- Bulk Density:900 kg/m3
- Boiling Point:400 °C (anhydrous substance),(decomposition)
- Vapor Pressure:(20 °C)
- Flash Point:250 °C (anhydrous substance)
- Density:1.42 g/cm3 (20 °C)
- pH:8 - 9.5 (10 g/l, H2O, 20 °C)
- Ignition Point:607 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06267.0500_TDS_EN MERC1.06267.1000_TDS_EN ProductPage_106267
Product specification
msds_106267
SDS
Frequently Bought Together