Sodium pyruvate solution 100 mM cell culture reagent
Catalog # LONZBEBP13-115E
Supplier: LONZA
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
:
:110.04 g/mol
Storage Temperature:Refrigerator
EINECS:204-024-4
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586