Charcoal activated Ph. Eur., SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Carbon activated, Activated carbon, Activated charcoal
CAS Number:7440-44-0
:64365-11-3
Formula:C
MDL Number:MFCD00133992
Density:2 g/cm³ (20 °C)
Molecular Weight:12.01 g/mol
Melting Pt:3550
Merck Index:13,01818
Storage Temperature:Ambient
EINECS:231-153-3
UN:1362
REACH:01-2119488894-16
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together