Anti-Nkx2.5 Polyclonal Antibody
Catalog # BSBTPB9293
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Nkx2.5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for NKX2.5/NKX2-5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. NKX2.5/NKX2-5 information: Molecular Weight: 34918 MW; Subcellular Localization: Nucleus ; Tissue Specificity: Expressed only in the heart.
Type: Primary
Antigen: Nkx2.5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse