Anti-IGF1 Goat Polyclonal Antibody (Biotin)
Catalog # PRSIXP-5159BT
Supplier: ProSci Inc.
Specifications
- Antigen name:Insulin Like Growth Factor 1
- Antigen symbol:IGF1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Mechano growth factor|Insulin-like growth factor I|IGF-I|MGF|Somatomedin-C|IGFI
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant mIGF-I (murine IGF-I).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Anti-mIGF-I specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IGF1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse