Anti-APOL1 Goat Polyclonal Antibody
Catalog # PRSI46-746
Supplier: ProSci Inc.
Specifications
- Antigen name:Apolipoprotein L1
- Antigen symbol:APOL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:APOL|ApoL-I|Apolipoprotein L-I|APO-L|Apolipoprotein L1|FSGS4|APOL-I
- Concentration:500 ug/mL
- Immunogen:ApoL1 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of ApoL1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 45 kDa band observed in human frontal cortex lysates (calculated MW of 44.0 kDa according to NP_003652.2;). Recommended concentration: 1-3 μg/mL.
ApoL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ApoL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human