Order Entry
Netherlands
ContactUsLinkComponent
Anti-TPD52 Goat Polyclonal Antibody
Anti-TPD52 Goat Polyclonal Antibody
Catalog # PRSI46-506
Supplier:  ProSci Inc.
undefined
Anti-TPD52 Goat Polyclonal Antibody
Catalog # PRSI46-506
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tumor Protein D52
  • Antigen symbol:
    TPD52
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    N8L|hD52|tumor protein D52|PC-1|PC1/MRPS28 fusion protein|D52|prostate and colon associated protein|TPD52|PrLZ
  • Concentration:
    500 ug/mL
  • Immunogen:
    TPD52 antibody was raised against an 11 amino acid synthetic peptide near the C-Terminus of TPD52.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

TPD52 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TPD52 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TPD52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human