- Antigen name:Exoribonuclease 1
- Antigen symbol:ERI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:protei|enhanced RNAi three prime mRNA exonuclease homolog 1|Eri-1 homolog|3'-5' exoribonuclease 1|histone mRNA 3'-exonuclease 1|3'HEXO|histone mRNA 3'-end-specific exoribonuclease|histone mRNA 3' end-specific exonuclease|HEXO|THEX1|3' exoribonuclease|3'-5' exonuclease ERI1
- Immunogen:A synthetic peptide corresponding a region of human THEX1.
- Size:50 µg
- Pk:50 µG
THEX1 contains 1 SAP domain and 1 exonuclease domain. It is an RNA exonuclease that binds to the 3' end of histone mRNAs and probably degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. It is also able to degrade the 3' overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ERI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human