Lens cleaning tissues
Supplier: Ahlstrom-Munksjö
Certificates
About this item
These very fine lens cleaning tissues are made of pure manila vegetable fibre.
- Non abrasive
Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.
12 g/m²
Frequently Bought Together