Anti-RAPGEF2 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-11581R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Rap Guanine Nucleotide Exchange Factor 2
- Antigen symbol:RAPGEF2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Neural RAP guanine nucleotide exchange protein|PDZ-GEF1|PDZ GEF1|Rapgef2|NRAPGEP|PDZGEF1|RA-GEF|KIAA0313|RA GEF|RPGF2_HUMAN.|PDZ domain containing guanine nucleotide exchange factor 1|PDZ domain-containing guanine nucleotide exchange factor 1|Rap guanine nucleotide exchange factor 2|nRap GEP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
PDZ-GEF1 is a 1,499 amino acid cell membrane protein that functions as a guanine nucleotide exchange factor for Rap 1A, Rap 1B and Rap 2B GTPases. Expressed at highest levels in brain, PDZ-GEF1 is found at low levels in placenta, heart, lung and kidney, and undergoes post-translational phosphorylation following DNA damage. PDZ-GEF1 interacts with MAGI-2 and contains one Ras-GEF domain, a Ras-associating domain, one PDZ (DHR) domain, a single N-terminal Ras-GEF domain and a cyclic nucleotide-binding domain. The gene encoding PDZ-GEF1 maps to human chromosome 4q32.1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAPGEF2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat