Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-EP2 Rabbit Polyclonal Antibody [clone: Q299]
  BWRLBS2598
Anti-EP2 Rabbit Polyclonal Antibody [clone: Q299]
  BWRLBS2598

 

  • Antibody type:
    Primary
  • Antigen symbol:
    EP2
  • Clonality:
    Polyclonal
  • Clone:
    Q299
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 280-330 of Human EP2.

EP2 (Q299) pAb detects endogenous levels of EP2 protein.

Type: Primary
Antigen: EP2
Clonality: Polyclonal
Clone: Q299
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat