Ferrules for Thermo Scientific GCs
Supplier: Restek
Certificates
About this item
Graphite Ferrules for Vu2 union connectors.
- High-purity, high-density graphite
- Smoother surface and cleaner edges than conventional graphite ferrules
- Contain no binders that can off -gas or adsorb analytes
- Stable to 450 °C
Frequently Bought Together