Rubella rubella grade IV antigen (strain HPV77) >90% viral protein, PBS (from Vero cells)
Catalogus # BIRBORB81837-1
Leverancier: Biorbyt
CAS-nummer:
Specificaties
- Pk:1 mg
- Source:Vero cells
- Species:Rubella
- Environmentally Preferable:
- Protein synonyms:Rubella Grade IV Antigen (Strain HPV77) >90% Viral
- Protein/peptide name:Rubella Grade IV Antigen (Strain HPV77) >90% Viral Protein, PBS
- Purity:>90%
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB
- Purification:Purified