Order Entry
Netherlands
ContactUsLinkComponent
Anti-MYH11 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MYH11/923]
Anti-MYH11 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MYH11/923]
Catalog # BNUM0923-50
Supplier:  Biotium
Anti-MYH11 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MYH11/923]
Catalog # BNUM0923-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myosin Smooth Muscle Heavy Chain
  • Antigen symbol:
    MYH11
  • Clonality:
    Monoclonal
  • Clone:
    Clone MYH11/923
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat
  • Environmentally Preferable:
  • Gene ID:
    460109
  • Antigen synonyms:
    SMMHC|AAT4|Myosin heavy chain 11 smooth muscle|SM2|Smooth muscle myosin heavy chain 11|Myosin-11|Myosin heavy chain 11 (MYH11)|SM1|SMHC
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors.

Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: MYH11/923
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat