Order Entry
Netherlands
ContactUsLinkComponent
Anti-FGF23 Mouse Monoclonal Antibody (CF647) [clone: FGF23/638]
Anti-FGF23 Mouse Monoclonal Antibody (CF647) [clone: FGF23/638]
Supplier:  Biotium
Certificates
Anti-FGF23 Mouse Monoclonal Antibody (CF647) [clone: FGF23/638]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470638-100
  • BNC470638-500
  • Antigen symbol:
  • FGF23
  • FGF23
  • Antigen name:
  • Fibroblast growth factor 23
  • Fibroblast growth factor 23
  • Antigen synonyms:
  • PHPTC|Phosphatonin|Tumor-derived hypophosphatemia-inducing factor|Fibroblast growth factor 23|ADHR|HPDR2|FGFN|FGF-23|HYPF
  • PHPTC|Phosphatonin|Tumor-derived hypophosphatemia-inducing factor|Fibroblast growth factor 23|ADHR|HPDR2|FGFN|FGF-23|HYPF
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone FGF23/638
  • Clone FGF23/638
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 287370
  • 287370
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together