Order Entry
Netherlands
ContactUsLinkComponent
Anti-IL2RA Mouse Monoclonal Antibody (CF640R) [clone: IL2RA/423]
Anti-IL2RA Mouse Monoclonal Antibody (CF640R) [clone: IL2RA/423]
Supplier:  Biotium
Certificates
Anti-IL2RA Mouse Monoclonal Antibody (CF640R) [clone: IL2RA/423]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400423-100
  • BNC400423-500
  • Antigen symbol:
  • IL2RA
  • IL2RA
  • Antigen name:
  • CD25
  • CD25
  • Antigen synonyms:
  • IL2R alpha chain|T Cell Growth Factor Receptor|TCGFR|Ly43|p55|IL2 Receptor alpha|p55 chain|IL2R|TAC antigen|Interleukin-2 receptor subunit alpha|IL2R subunit alpha IL2RA
  • IL2R alpha chain|T Cell Growth Factor Receptor|TCGFR|Ly43|p55|IL2 Receptor alpha|p55 chain|IL2R|TAC antigen|Interleukin-2 receptor subunit alpha|IL2R subunit alpha IL2RA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone IL2RA/423
  • Clone IL2RA/423
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 231367
  • 231367
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together