(2-Bromoethyl)cyclohexane 98%
: 2-Cyclohexylethyl bromide, 1-Bromo-2-cyclohexylethane
:1647-26-3
:
:C₈H₁₅Br
:MFCD00019398
:209 °C (760 mmHg)
:–57 °C
:1.221 g/cm³
:191.11 g/mol
:96 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4910 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together