Methyl-1H-imidazole-5-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl 4-imidazolecarboxylate, Methyl 1H-imidazole-5-carboxylate
CAS Number:17325-26-7
:
Formula:C₅H₆N₂O₂
MDL Number:MFCD00216584
Molecular Weight:126.11 g/mol
Melting Pt:154…156 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together