Ethylene diacrylate 95% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethylene glycol diacrylate
CAS Number:2274-11-5
:
Formula:C₈H₁₀O₄
MDL Number:MFCD00008629
Boiling Pt:62…65 °C (2 mmHg)
Density:1.04 g/cm³
Molecular Weight:170.164 g/mol
Flash Pt:101 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1 III
:GHS06
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4520 to 1.4560 (20°C, 589 nm)
- GC:≥94 %
- Stabilizer:100 to 400 ppm MEHQ
Specification Test Results
Frequently Bought Together