Succinamic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:638-32-4
:
Formula:C₄H₇NO₃
MDL Number:MFCD00008041
Molecular Weight:117.104 g/mol
Melting Pt:153…157 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:153 to 157 °C
- Titration with NaOH:≥96.0 %
- Ammonium (NH4):≤0.003 %
- Iron (Fe):≤0.003 %
Specification Test Results
Frequently Bought Together