Leuco malachite green, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-(Phenylmethylene)bis(N,N-dimethylaniline), 4,4'-Tetramethyldiaminotriphenylmethane, 4,4'-Benzylidenebis(N,N-dimethylaniline), Leucomalachite green
CAS Number:129-73-7
:
Formula:C₂₃H₂₆N₂
MDL Number:MFCD00008315
Molecular Weight:330.48 g/mol
Melting Pt:100…102 °C
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light green to light brown to light blue Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- TMS:0.03 %
Specification Test Results
Frequently Bought Together