(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol 96%
: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
:2964-48-9
:
:C₉H₁₂N₂O₄
:MFCD00007359
:212.2026 g/mol
:160…166 °C
:Ambient
- Environmentally Preferable:
- Appearance:Yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:160 to 166 °C
- Specific optical rotation:+29° to +33° (20 °C, 589 nm) (c=1, 6 N HCl)
- Titration with HClO4:≥95 %
Frequently Bought Together