Yeast nitrogen base, without amino acids, without ammonium sulfate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Yeast nitrogen base without amino acids and ammonium sulfate is used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate. Addition of nitrogen and carbon sources is required.
MDL No.: MFCD00217952
Specifications
- Environmentally Preferable:
Specifications