meso-Hydrobenzoin 95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 579-43-1 Molecular Formula: C14H14O2 Formula Weight: 214.27 Melting Point: 134-136° Beilstein Reference: 2050814 MDL No.: MFCD00064253
Synonyms: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol
- CAS Number:579-43-1
- Formula:C₁₄H₁₄O₂
- MDL Number:MFCD00064253
- Molecular Weight:214.26 g/mol
- Merck Index:14,4777
- Melting Pt:134…136 °C
- Storage Temperature:Kamertemperatuur
- EINECS:207-758-3
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 579-43-1 Molecular Formula: C14H14O2 Formula Weight: 214.27 Melting Point: 134-136° Beilstein Reference: 2050814 MDL No.: MFCD00064253
Synonyms: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol
- CAS Number:579-43-1
- Formula:C₁₄H₁₄O₂
- MDL Number:MFCD00064253
- Molecular Weight:214.26 g/mol
- Merck Index:14,4777
- Melting Pt:134…136 °C
- Storage Temperature:Kamertemperatuur
- EINECS:207-758-3
Documents
Product Family Options
Product Information
- VPEAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



