Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(styrene)
Poly(styrene) 10% (w/w) dispersion in water, latex microsphere 1 micron
STYRENE 10% (w/w) dispersion in water, latex microsphere 1 micron
41917.7C
:
Thermo Fisher Scientific
:
STYRENE 10% (w/w) dispersion in water, latex microsphere 1 micron
41917.7C
:
Thermo Fisher Scientific
:
Pk:
15 mL
Environmentally Preferable:
:
:
125000 - 250000 g/mol
:
Ambient
:
9003-53-6
:
C₈H₈
:
MFCD00084450