Yttrium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38245-39-5
:
Formula:Y₂(CO₃)₃·xH₂O
MDL Number:MFCD00149943
Molecular Weight:357.84 g/mol (anhy)
Merck Index:14,10107
Storage Temperature:Ambient
EINECS:209-119-4
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together