Peqlab peqGOLD, Agarose, Universal
Supplier: VWR Chemicals
Certificates
About this item
High quality standard agarose for routine analysis with broad separation range (0,05 to 50 kbp). Ideal for blotting applications due to high gel strength. Free from RNases, DNases and enzyme inhibitors.
Related Information
Certificates
Search by Lot Number35-10XX_e_v1116_m 35-10XX_v1016_m
Datasheet
peqGOLD Universal-Agarose VWR® Products for Electrophoresis
Product Literature
spec_PEQL35-1020 spec_VWRC35-1010 spec_VWRC35-1010 spec_VWRC35-1020 spec_VWRC35-1020
Product specification
Frequently Bought Together