Anti-CRP Mouse Monoclonal Antibody [clone: 5A9]
Catalog # ABNOMAB1089
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:C-Reactive Protein
- Antigen symbol:CRP
- Clonality:Monoclonal
- Clone:5A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:1401
- Antigen synonyms:CRP1|MGC149895|C-reactive protein|pentraxin 1|CRP|D1S181E|RP11-419N10.4|CSRP|CYRP|MGC88244|PTX1
- Amino acid number:19 to 224
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 19-224 of human CRP.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CRP.
Recommended Dilutions: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: 5A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse