Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]
ABNOMAB1437
: Abnova
- Antibody type:Primary
- Antigen name:Cell division cycle 6
- Antigen symbol:CDC6
- Clonality:Monoclonal
- Clone:IMD-6
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:990
- Antigen synonyms:HsCDC18|Hscell division cycle 6|CDC18L
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human CDC6.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant CDC6.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone: IMD-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human