Order Entry
Netherlands
ContactUsLinkComponent
Anti-MCL1 Mouse Monoclonal Antibody [clone: MCL.C9]
Anti-MCL1 Mouse Monoclonal Antibody [clone: MCL.C9]
Catalog # ABNOMAB8825
Supplier:  Abnova
Anti-MCL1 Mouse Monoclonal Antibody [clone: MCL.C9]
Catalog # ABNOMAB8825
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Induced myeloid leukemia cell differentiation protein
  • Antigen symbol:
    MCL1
  • Clonality:
    Monoclonal
  • Clone:
    MCL.C9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4170
  • Antigen synonyms:
    BCL2L3|MCL1L|Mcl-1|MCL1S|MGC104264|TM|MGC1839|EAT
  • Storage buffer:
    PBS, pH 7,4 (0,05% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human MCL1.
  • Purification:
    Protein A purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant MCL1.

Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: MCL.C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human